Fortfahren mit
26208
Hamilton® Spritze, Serie 1000 GASTIGHT®, PTFE-Luer-Lock
1001TLL, PTFE Luer lock (with slots), volume 1 mL, needle size (not included)
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
PackungsgrößeSKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41122001
NACRES:
NB.03
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
PTFE Luer lock (with slots)
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81327
needle L
, n/a
volume
1 mL
fitting
luer lock (without slots)
compatibility
for use with Hamilton 1001TLL (PTFE Luer lock)
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Application
It has been used in an experiment performed to inject magnetic beads into Lab-on-a-Chip system in order to demonstrate controlled movement of the magnetic beads within a microfluidic channel. It was also used in alkene termination through olefin cross-metathesis reaction performed with gallium nitride wafers.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
209972099921967
material
PTFE Luer lock (with slots)
material
PTFE Luer lock
material
PTFE Luer lock
material
Kel-F CTFE Luer lock
fitting
luer lock (without slots)
fitting
luer lock (without slots)
fitting
luer lock (without slots)
fitting
luer lock (without slots)
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
needle L
, n/a
needle L
, n/a
needle L
, n/a
needle L
, n/a
compatibility
for use with Hamilton 1001TLL (PTFE Luer lock)
compatibility
for use with Hamilton 1001TLL (PTFE Luer lock)
compatibility
for use with purge and trap (US EPA)
compatibility
for use with Hamilton 1100TLL (PTFE Luer lock)
volume
1 mL
volume
1 mL
volume
5 mL
volume
100 mL
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Controlled counter-flow motion of magnetic bead chains rolling along microchannels.
Karle, Marc, et al.
Microfluidics and nanofluidics, 10.4, 935-939 (2011)
Pharmacology for the Prehospital Professional, 54-54 (2009)
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Global Trade Item Number
SKUGTIN
2620804061832873923