Größe auswählen
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
Kel-F CTFE Luer lock
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 86020
needle L
, n/a
volume
100 mL
fitting
luer lock (without slots)
compatibility
for use with Hamilton 1100TLL (PTFE Luer lock)
General description
Application
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material Kel-F CTFE Luer lock | material PTFE Luer lock | material PTFE Luer lock | material PTFE Luer lock (with slots) |
| fitting luer lock (without slots) | fitting luer lock (without slots) | fitting luer lock (without slots) | fitting luer lock (without slots) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| needle L , n/a | needle L , n/a | needle L , n/a | needle L , n/a |
| needle size (not included) | needle size (not included) | needle size (not included) | needle size (not included) |
| compatibility for use with Hamilton 1100TLL (PTFE Luer lock) | compatibility for use with Hamilton 1001TLL (PTFE Luer lock) | compatibility for use with purge and trap (US EPA) | compatibility for use with Hamilton 1001TLL (PTFE Luer lock) |
Still not finding the right product?
Explore all of our products under Hamilton® Spritze, Serie 1000 GASTIGHT®, PTFE-Luer-Lock
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 21967 | 04061832873473 |