Ansicht ändern
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
UNSPSC Code:
41122001
NACRES:
NB.03
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
Kel-F CTFE Luer lock
needle size
22 gauge (not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 85020
needle L
, n/a
volume
50 mL
fitting
luer lock (without slots)
General description
Hamilton® syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material Kel-F CTFE Luer lock | material Kel-F CTFE Luer lock | material PTFE Luer lock | material Kel-F CTFE Luer lock |
| fitting luer lock (without slots) | fitting - | fitting luer lock (without slots) | fitting luer lock (without slots) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| manufacturer/tradename Hamilton 85020 | manufacturer/tradename Hamilton 82520 | manufacturer/tradename Hamilton 81420 | manufacturer/tradename Hamilton 86020 |
| volume 50 mL | volume 25 mL | volume 2.5 mL | volume 100 mL |
| needle size 22 gauge (not included) | needle size 22 gauge (not included) | needle size (not included) | needle size (not included) |
Still not finding the right product?
Explore all of our products under Hamilton® Spritze, Serie 1000 GASTIGHT®, PTFE-Luer-Lock
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192 -192 (2012)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 20707 | 04061832873107 |