Größe auswählen
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|---|---|---|
| 1 each | Warenkorb auf Verfügbarkeit prüfen | € 211,00 |
Über diesen Artikel
material
stainless steel clamp(s)
feature
autoclavable
manufacturer/tradename
MAS-100 Atmos®
application(s)
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 Atmos®
General description
Place the silicone gasket (ref. 1.17364) on top of the closed sampling head and fix the pressure connection tubing with the 34 mm tri-clamp
Application
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| compatibility for use with MAS-100 Atmos® | compatibility for use with MAS-100® Iso | compatibility for use with MAS-100 Atmos® | compatibility for use with MAS-100 Atmos® |
| material stainless steel clamp(s) | material stainless steel | material silicone gasket | material PTFE tube |
| application(s) compressed gas monitoring | application(s) air monitoring | application(s) compressed gas monitoring | application(s) compressed gas monitoring |
| manufacturer/tradename MAS-100 Atmos® | manufacturer/tradename - | manufacturer/tradename MAS-100 Atmos® | manufacturer/tradename MAS-100 Atmos® |
| feature autoclavable | feature installed | feature autoclavable | feature autoclavable |
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1173670001 | 04065267770734 |