Größe auswählen
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|---|---|---|
| 1 ea | Warenkorb auf Verfügbarkeit prüfen | € 118,00 |
Über diesen Artikel
€ 118,00
material
borosilicate glass flask, round bottom , single-neck (high vaccum valve)
joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F541950
capacity
50 mL, 50 mL
valve size
0-4 mm
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material borosilicate glass flask, single-neck (high vaccum valve), round bottom | material borosilicate glass flask, round bottom , single-neck (high vaccum valve) | material borosilicate glass flask, round bottom , single-neck (high vaccum valve) | material borosilicate glass flask, round bottom , single-neck (high vaccum valve) |
| manufacturer/tradename Synthware F541950 | manufacturer/tradename Synthware F541050 | manufacturer/tradename Synthware F549100 | manufacturer/tradename Synthware F541100 |
| capacity 50 mL , 50 mL | capacity 50 mL , 50 mL | capacity 100 mL , 100 mL | capacity 100 mL , 100 mL |
| joint joint: ST/NS19/22 | joint joint: ST/NS14/20 | joint joint: ST/NS19/22 | joint joint: ST/NS14/20 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| valve size 0-4 mm | valve size 0-4 mm | valve size 0-4 mm | valve size 0-4 mm |
Still not finding the right product?
Explore all of our products under Synthware™ reaction flask with high vacuum valve
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SYNF541950-1EA | 04061832195049 |