Größe auswählen
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|---|---|---|
| 12 ea | Warenkorb auf Verfügbarkeit prüfen | € 163,00 |
Über diesen Artikel
€ 163,00
material
acrylonitrile-butadiene-styrene (ABS) , yellow
feature
Holds 50 x slides 76x26 mm
packaging
pack of 12 ea
manufacturer/tradename
Ted Pella 2304-5-CS
L × W × H
164 mm (6.46 in.) × 110 mm (4.33 in.) × 32 mm (1.26 in.)
General description
Features and Benefits
- Numbered index sheet in lid for quick identification
- Foam liner
- Stackable
- ABS-durable/impact resistant
- Hinged lid
- Unique sliding compression latch
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| manufacturer/tradename Ted Pella 2304-5-CS | manufacturer/tradename Ted Pella 2302-5-CS | manufacturer/tradename Ted Pella 2306-5-CS | manufacturer/tradename Ted Pella 2304-6-CS |
| material acrylonitrile-butadiene-styrene (ABS) , yellow | material acrylonitrile-butadiene-styrene (ABS) , yellow | material acrylonitrile-butadiene-styrene (ABS) , yellow | material acrylonitrile-butadiene-styrene (ABS) , black |
| packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 12 ea |
| L × W × H 164 mm (6.46 in.) × 110 mm (4.33 in.) × 32 mm (1.26 in.) | L × W × H 209 mm (8.23 in.) × 172 mm (6.77 in.) × 32 mm (1.26 in.) | L × W × H 104 mm (4.10 in.) × 94 mm (3.70 in.) × 32 mm (1.26 in.) | L × W × H 164 mm (6.46 in.) × 110 mm (4.33 in.) × 32 mm (1.26 in.) |
| feature Holds 50 x slides 76x26 mm | feature Holds 100 x slides 76x26 mm | feature Holds 20 x slides 76x26 mm | feature Holds 50 x slides 76x26 mm |
Still not finding the right product?
Explore all of our products under PELCO® Sliding Latch Microslide Storage Boxes
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| Z742402-12EA | 04061833678510 |