Über diesen Artikel
NACRES:
JA.01
UNSPSC Code:
12352200
material
polyethylene , silver (drawers)
manufacturer/tradename
TrippNT 51139
W × H × D
24 1/2 × 3 3/8 × 15 1/2 in., drawers
weight
67 lb
General description
An extra-wide compact cart with 4 drawers; fits under most counters, tables, desks and work benches.
- Massive under counter storage
- One piece construction
- Non metal rust proof polymer construction
- Decontaminates with bleach or aqueous solutions
- Built-in tray top and handle for push pull control
- White polyethylene body, ABS drawers
- 13 colors
- 4 full-extension drawers
- Drawer headspace: 4 1/2 inches
- 3 inch polypropylene swivel casters, 2 locking
- American Made
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?