Ansicht ändern
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
Lineare Formel:
H2NC6H4CONHCH2CH2N(C2H5)2·HCl
CAS-Nummer:
Molekulargewicht:
271.79
EC Number:
210-381-7
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
3729517
MDL number:
Form:
powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenform
powder
mp
167-169 °C (lit.)
solubility
H2O: soluble, ethanol: soluble
originator
Bristol-Myers Squibb
SMILES string
Cl.CCN(CC)CCNC(=O)c1ccc(N)cc1
InChI
1S/C13H21N3O.ClH/c1-3-16(4-2)10-9-15-13(17)11-5-7-12(14)8-6-11;/h5-8H,3-4,9-10,14H2,1-2H3,(H,15,17);1H
InChI key
ABTXGJFUQRCPNH-UHFFFAOYSA-N
Gene Information
Biochem/physiol Actions
Inhibits DNA methyltransferase and modulates epigenetic regulation of gene expression. Na+ channel blocker and Class IA anti-arrhythmic.
Features and Benefits
This compound is a featured product for ADME Tox and Gene Regulation research. Discover more featured ADME Tox and Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.
Still not finding the right product?
Explore all of our products under Procainamid -hydrochlorid
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Lagerklasse
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Y H Kim et al.
Circulation, 100(6), 666-674 (1999-08-10)
Ventricular fibrillation (VF) is maintained by 2 mechanisms: first by reentry formation and second by spontaneous wave break or wave splitting. We hypothesized that spontaneous wave break results from a critical shortening of the action potential duration (APD) during VF
Ian G Stiell et al.
Annals of emergency medicine, 57(1), 13-21 (2010-09-25)
Although recent-onset atrial fibrillation and flutter are common arrhythmias managed in the emergency department (ED), there is insufficient evidence to help physicians choose between 2 competing treatment strategies, rate control and rhythm control. We seek to evaluate variation in ED
Z Wojnarowska et al.
The Journal of chemical physics, 136(16), 164507-164507 (2012-05-09)
The pharmaceuticals, procaine hydrochloride and procainamide hydrochloride, are glass-forming as well as ionically conducting materials. We have made dielectric measurements at ambient and elevated pressures to characterize the dynamics of the ion conductivity relaxation in these pharmaceuticals, and calorimetric measurements