Größe auswählen
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|---|---|---|
| 10 ea | Warenkorb auf Verfügbarkeit prüfen | € 76,50 |
Über diesen Artikel
€ 76,50
material
colorless , colorless wells
well size
384
sterility
non-sterile
feature
skirt, Forensic DNA Grade, skirted
packaging
pack of 10 (individually wrapped)
manufacturer/tradename
Eppendorf® 0030129628 (Global)
technique(s)
DNA amplification: suitable (Forensic grade)
size
384 wells
working volume
40 μL
General description
Features and Benefits
- Optimized rim for barcodes and automated work processes
- Stackable
- Within SBS footprint recommendations
- Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler X50 and Mastercycler nexus
- 40 µL max. well volume A
- Ideal for quantitative real-time PCR as well – OptiTrack matrix for faster sample identification and fewer pipetting errors
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| sterility non-sterile | sterility - | sterility - | sterility non-sterile |
| feature skirt, skirted, Forensic DNA Grade | feature skirt, Forensic DNA Grade, Skirt, Skirted | feature skirt, Forensic DNA Grade, Semi-Skirted, Skirt (Semi skirted) | feature skirt, Forensic DNA Grade, Skirted |
| manufacturer/tradename Eppendorf® 0030129628 (Global) | manufacturer/tradename Eppendorf® 0030129601 (Global) | manufacturer/tradename Eppendorf® 0030129610 (Global) | manufacturer/tradename Eppendorf® 0030129636 (Global) |
| material colorless , colorless wells | material colorless , colorless wells | material colorless , colorless wells | material white plate, white wells |
| working volume 40 μL | working volume 150 μL | working volume 250 μL | working volume 150 μL |
| size 384 wells | size 96 wells | size 96 wells | size 96 wells |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| EP0030129628-10EA | 04061835081769 |