description
Micromatic 8 channel aspirator
feature
autoclavable
manufacturer/tradename
Corning 3389
General description
Helps safely vacuum aspirate medium or buffers from the apical portion of the HTS Transwell-96 inserts. Using the 8-Channel Aspirator reduces cell death or detachment caused by too rapid removal of solutions or media during vacuum aspiration.
Legal Information
Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?