Fortfahren mit
41041
L-Fucono-1,4-lacton
≥90% (TLC)
Synonym(e):
6-Deoxy-L-galactonsäure-γ-lacton, L-Fuconsäure-γ-lacton
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
Empirische Formel (Hill-System):
C6H10O5
CAS-Nummer:
Molekulargewicht:
162.14
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
246-121-4
Beilstein/REAXYS Number:
82272
Assay:
≥90% (TLC)
Form:
powder
assay
≥90% (TLC)
form
powder
optical activity
[α]/D +78±3°, c = 2 in H2O
suitability
corresponds for H-NMR
storage temp.
2-8°C
SMILES string
O[C@H]([C@@H]([C@@H](O)C)O1)[C@H](O)C1=O
InChI
1S/C6H10O5/c1-2(7)5-3(8)4(9)6(10)11-5/h2-5,7-9H,1H3/t2-,3?,4+,5+/m0/s1
InChI key
VASLEPDZAKCNJX-MEFNJWCVSA-N
Biochem/physiol Actions
Metabolite in the fructose and mannose metabolism
Lagerklasse
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Isolation of diphosphopyridine nucleotide-dependent L-fucose dehydrogenase from pork liver.
H Schachter et al.
The Journal of biological chemistry, 244(17), 4785-4792 (1969-09-10)
H S Bleeg et al.
European journal of biochemistry, 127(2), 391-396 (1982-10-01)
An enzyme from Saccharomyces cerevisiae which catalyzes the reaction: L-galactonolactone + O2 leads to L-ascorbate + H2O2 has been purified 466-fold from the mitochondrial fraction of a yeast homogenate. The enzyme has several properties that are different from the L-galactonolactone
Merlin Eric Hobbs et al.
Biochemistry, 53(28), 4661-4670 (2014-06-26)
A previously unknown metabolic pathway for the utilization of l-galactose was discovered in a prevalent gut bacterium, Bacteroides vulgatus. The new pathway consists of three previously uncharacterized enzymes that were found to be responsible for the conversion of l-galactose to
Global Trade Item Number
SKUGTIN
41041-100MG04061831579949
41041-500MG04061831579956