Fortfahren mit
04971
Atto 680 Maleimid
BioReagent, suitable for fluorescence, ≥90% (HPLC)
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32
MDL number:
product line
BioReagent
assay
≥90% (HPLC)
manufacturer/tradename
ATTO-TEC GmbH
solubility
DMF: soluble, H2O: soluble (pH 7.0), ethanol: soluble, methanol: soluble
λ
in acetonitrile: water (30/70; with 0.1% trifluoroacetic acid)
UV absorption
λ: 683-689 nm Amax
suitability
corresponds for coupling to thiols, suitable for fluorescence
storage temp.
−20°C
InChI
1S/C33H37N5O7S/c1-4-36-12-5-7-21-15-24-28(17-26(21)36)45-29-18-27-23(16-25(29)35-24)22(20-46(42,43)44)19-33(2,3)38(27)13-6-8-30(39)34-11-14-37-31(40)9-10-32(37)41/h9-10,15-19H,4-8,11-14,20H2,1-3H3,(H-,34,39,42,43,44)
InChI key
DRRZFCNOIUXBKW-UHFFFAOYSA-N
Application
Atto 680 maleimide can be used for the fluorescence labeling of biological molecules (i.e., DNA, RNA, and proteins) with high molecular absorption and quantum yeild, and good thermal and photo-stability. A zwitterionic dye carrying a net electrical charge of zero.
Legal Information
This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.
Lagerklasse
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Slawomir Gwiazda et al.
Biochimie, 94(7), 1457-1463 (2012-04-03)
The RNA-world-theory is one possible explanation of how life on earth has evolved. In this context it is of high interest to search for molecular systems, capable of self-organization into structures with increasing complexity. We have engineered a simple catalytic
The design of a peptide linker group to enhance the SERS signal intensity of an atto680 dye-nanoparticle system.
Drake, P., et al.
Journal of Raman Spectroscopy, 41, 1248-1253 (2010)
Irene Drude et al.
Biochemical and biophysical research communications, 363(1), 24-29 (2007-09-11)
Over the past two decades, the structure and mechanism of catalytic RNA have been extensively studied; now ribozymes are understood well enough to turn them into useful tools. After we have demonstrated the twin ribozyme mediated insertion of additional nucleotides
Global Trade Item Number
SKUGTIN
301841-1G04061826665572
04971-1MG-F04061826670316