Fortfahren mit
Y.11086
Prove Connect to LIMS
Unlimited License
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
12352200
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
General description
In many cases it is advisable to export results and measurement data sets, e.g. to archive them or to analyze them in greater detail using a suitable software tool. Spectroquant® Prove Connect to LIMS is a desktop application to connect your Spectroquant® Prove instruments to laboratory software systems such as LIMS and ELNs. The software receives meisasurement data from Spectroquant® Prove instruments over the local network and provides it to laboratory information systems.
Spectroquant® Prove Connect runs as a background service and supports the following file formats:
−XML
−CSV
This allows an easy integration of Spectroquant® Prove instruments into an existing laboratory IT infrastructure.
This item can only be used in combination with Spectroquant® Prove spectrophotometers. A PC with Microsoft Windows 7 SP1, Windows 8, or Windows 10 and 1 GB of free disk space, 4 GB of RAM, and Network Connection (LAN) is mandatory to ensure compatibility.
Legal Information
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany