Fortfahren mit
NA25/25
Diaphragm for NOVASEPTIC® Valve
PTFE
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
PackungsgrößeSKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
31411800
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
Quality Level
material
PTFE , PTFE device (polytetrafluoroethylene)
feature
hydrophobic (Surface)
parameter
-40-170 °C temp. range (Continous use), -40-250 °C temp. range (Short term use)
weight
0.04 kg (0.088 lb)
Application
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Packaging
The Diaphragm is packaged in a closed box.
Other Notes
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.2600 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
NA12/22NA76/25NA12/25
material
PTFE , PTFE device (polytetrafluoroethylene)
material
EPDM rubber device (ethylene propylene diene monomer rubber), PTFE
material
PTFE , PTFE device (polytetrafluoroethylene)
material
PTFE , PTFE device (polytetrafluoroethylene), valve (Diaphragm, PTFE)
weight
0.04 kg (0.088 lb)
weight
0.01 kg (0.022 lb)
weight
0.16 kg (0.353 lb)
weight
0.01 kg (0.022 lb)
feature
hydrophobic (Surface)
feature
hydrophobic (Surface)
feature
hydrophobic (Surface)
feature
hydrophobic (Surface)
parameter
-40-170 °C temp. range (Continous use), -40-250 °C temp. range (Short term use)
parameter
-30-140 °C temp. range (Short term use), -6-(-1) bar pressure range
parameter
-40-170 °C temp. range (Continuous use), -40-250 °C temp. range (Short term use)
parameter
-40-170 °C temp. range (Continous use), -40-250 °C temp. range (Short term use)
Quality Level
SP2
Quality Level
SP2
Quality Level
SP2
Quality Level
SP2
Still not finding the right product?
Explore all of our products under