Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
KU42.33C, monoclonal
Application:
ELISA, FACS, IHC, IP, WB
Citations:
-
conjugate
unconjugated
antibody form
purified antibody
clone
KU42.33C, monoclonal
purified by
using protein G
species reactivity
human, mouse
concentration
(Please refer to lot specific datasheet.)
technique(s)
ELISA: suitable, flow cytometry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG1κ
UniProt accession no.
target post-translational modification
unmodified
General description
BxPC-3 human pancreatic cancer cells.
~210 kDa observed; 161.70 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Application
Epitope: extracellular domain
Biochem/physiol Actions
Clone KU42.33C is a mouse monoclonal antibody that detects human CD109.
Physical form
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blotting in Panc-1 cell lysate.
Western Blotting Analysis: 2 µg/mL of this antibody detected CD109 in Panc-1 cell lysate.
Western Blotting Analysis: 2 µg/mL of this antibody detected CD109 in Panc-1 cell lysate.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Analysenzertifikate (COA)
Suchen Sie nach Analysenzertifikate (COA), indem Sie die Lot-/Chargennummer des Produkts eingeben. Lot- und Chargennummern sind auf dem Produktetikett hinter den Wörtern ‘Lot’ oder ‘Batch’ (Lot oder Charge) zu finden.
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MABS1928-100UG | 04061841039167 |
| MABS1928-25UG | 04061841039174 |