air monitoring cosmetics food and beverages pharmaceutical
compatibility
for use with MAS-100 Atmos®, for use with MAS-100 NT® (with filter)
General description
The HEPA H13 Filter 74 mm for MAS-100 NT/NT Ex is an replacement filter for MAS-100 NT® w. filter, MAS-100 NT® Ex w. filter as well as for MAS-100 NT® or MAS-100 NT® Ex which are upgraded with the mounting Kit for Filter. The filter should be replaced on yearly bases. To ease the workflow, it is rcommended to replace the filter along with the yearly calibration/adjustment of the microbial air sampler.
Application
The HEPA H13 Filter is a replacement filter for microbial air samplers MAS-100 NT® with filter and MAS-100 NT® Ex with filter. Microbial air sampling devices are extensively used in Pharmaceutical applications.
for use with MAS-100 Atmos®, for use with MAS-100 NT® (with filter)
compatibility
-
compatibility
-
compatibility
-
application(s)
air monitoring cosmetics food and beverages pharmaceutical
application(s)
-
application(s)
-
application(s)
-