Größe auswählen
| Packungsgröße | SKU | Verfügbarkeit | Preis |
|---|---|---|---|
| 1 ea | Warenkorb auf Verfügbarkeit prüfen | € 54,70 |
Über diesen Artikel
€ 54,70
material
borosilicate glass flask, round bottom , two-neck (angled side neck)
joint
center joint: ST/NS19/22, side joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F411925
capacity
25 mL, 25 mL
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material borosilicate glass flask, two-neck (angled side neck), round bottom | material borosilicate glass flask, round bottom , two-neck (angled side neck) | material borosilicate glass flask, round bottom , two-neck (angled side neck) | material borosilicate glass flask, round bottom , two-neck (angled side neck) |
| capacity 25 mL , 25 mL | capacity 25 mL , 25 mL | capacity 25 mL , 25 mL | capacity 250 mL , 250 mL |
| manufacturer/tradename Synthware F411925 | manufacturer/tradename Synthware F419925 | manufacturer/tradename Synthware F411125 | manufacturer/tradename Synthware F414251 |
| joint center joint: ST/NS19/22, side joint: ST/NS14/20 | joint center joint: ST/NS19/22, side joint: ST/NS19/22 | joint center joint: ST/NS14/20, side joint: ST/NS14/20 | joint center joint: ST/NS24/40, side joint: ST/NS14/20 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SYNF411925-1EA | 04061832193809 |