Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 464,00 |
Über diesen Artikel
€ 464,00
feature
incremental 0.02 mL, DE-M marking, with recirculation valve
manufacturer/tradename
BRAND 4600331
volume range
0.5-5 mL
fitting
(GL 28/S28), GL 32, GL 38, S 40
General description
Features and Benefits
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Other Notes
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| manufacturer/tradename BRAND 4600331 | manufacturer/tradename BRAND 4600321 | manufacturer/tradename BRAND 4600361 | manufacturer/tradename BRAND 4600341 |
| feature incremental 0.02 mL, with recirculation valve, DE-M marking | feature incremental 0.01 mL, DE-M marking, with recirculation valve | feature incremental 0.2 mL, DE-M marking, with recirculation valve | feature incremental 0.05 mL, DE-M marking, with recirculation valve |
| fitting (GL 28/S28), GL 38, GL 32, S 40 | fitting (GL 28/S28), GL 32, GL 38, S 40 | fitting (GL 32-33), GL 38, S 40 | fitting (GL 28/S28), GL 32, GL 38, S 40 |
| volume range 0.5-5 mL | volume range 0.2-2 mL | volume range 5-50 mL | volume range 1-10 mL |
Still not finding the right product?
Explore all of our products under BRAND® Dispensette® S Digital bottle-top dispenser
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?