MilliporeSigma
Passer à
B7410
Nalgene® bottles, style 2120
capacity 4,000 mL
Synonyme(s) :
HDPE bottles, nalgene bottle, nalgene bottles, Nalgene 2120-0010, laboratory container, vessel
Se connecter pour consulter les tarifs organisationnels et contractuels.
Changer de vue
ConditionnementRéférenceDisponibilitéPrix
1 ea
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
69,70 $
A propos de cet article
NACRES:
NB.17
UNSPSC Code:
41121800
69,70 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
material
high-density polyethylene bottle, polypropylene cap, polypropylene screw closure, round natural bottle
joint
Joints threaded neck (100)
description
Not autoclavable
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 2120-0010
bottle diam. × H
153 mm × 288 mm
capacity
4,000 mL
General description
Round, wide-mouth bottles
Features and Benefits
  • Versatile as it is chemical-resistant to acids, bases, and alcohols
  • Excellent for freezer storage up to −100°C (−148°F) for long-term purposes
  • Non-leakproof for liquids
  • Made from non-toxic materials
  • Linerless closures are recommended to prevent leaks, wrinkles, and contamination of reagents
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Comparer avec des articles similaires
Voir la comparaison complète
Montrer les différences
1 of 1
Cet article
B7285Z377333Z377325
material
high-density polyethylene bottle, polypropylene screw closure, polypropylene cap, round natural bottle
material
high-density polyethylene bottle, polypropylene cap, polypropylene screw closure, round bottle
material
polypropylene cap, polypropylene copolymer bottle (PPCO), square translucent bottle
material
polypropylene cap, polypropylene copolymer bottle (PPCO), square bottle, translucent bottle, wide-mouth bottle
feature
with cap
feature
with cap
feature
with cap
feature
autoclavable, with cap
manufacturer/tradename
Nalgene® 2120-0010
manufacturer/tradename
Nalgene® 2120-0005
manufacturer/tradename
Nalgene® 2110-0006
manufacturer/tradename
Nalgene® 2110-0002
capacity
4,000 mL
capacity
2,000 mL
capacity
175 mL
capacity
-
joint
Joints threaded neck (100)
joint
Joints threaded neck (100)
joint
Joints threaded neck (38)
joint
Joints threaded neck (28)
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pkg of 12
packaging
pkg of 12
Still not finding the right product?
Explore all of our products under Nalgene® bottles, style 2120
Numéro d'article de commerce international
RéférenceGTIN
B7410-1EA04061832613826