Saltar al contenido
MilliporeSigma

Saltar a

PLA0022

Rabbit anti-Rad21 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Sinónimos:

CDLS4, HR21, HRAD21, KIAA0078, MCD1, NXP-1, NXP1, RAD21 (S. pombe) homolog, RAD21 homolog, RAD21 homolog (S. pombe), SCC1, SCC1 homolog, hHR21, kleisin, nuclear matrix protein 1, protein involved in DNA double-strand break repair, sister chromatid cohesion 1

Iniciar sesión para ver los precios por organización y contrato.

Seleccione un Tamaño

Cambiar Vistas
Tamaño de envaseSKUDisponibilidadPrecio
100 μL
Comprobar disponibilidad del carrito
$540.00

Acerca de este artículo

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
4

$540.00


Comprobar disponibilidad del carrito

Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarle


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunoprecipitation (IP): 2-10 μg/mg, western blot: 1:2,000-1:10,000

accession no.

NP_006256.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... Rad21(5885)

General description

RAD21 (RAD21 homolog, S. pombe) is a novel nuclear protein located on chromosome 8q24.

Immunogen

The epitope recognized by PLA0022 maps to a region between residue 575 and the C-terminus (residue 631) human Rad21 homolog using the numbering given in entry NP_006256.1 (GeneID 5885).

Biochem/physiol Actions

RAD21 (RAD21 homolog, S. pombe) plays a crucial role in normal cell division. It acts as a main component of the cohesin complex for the proper chromosomal arrangement during cell division. The protein possesses two mitotic cleavage sites for the attachment of separase. It has been shown experimentally that at the beginning of anaphase stage, a caspase-like Esp1/separase attaches to the cleavage site of RAD21 and initiate sister chromatid separation, which is an essential step for the completion of cytokinesis. During double strand DNA damage repair, it helps the two DNA strands to adhere with each other. Mutation in RAD21 causes a congenital phenotype, cohesinopathy in humans characterized with growth retardation and minor skeletal anomalies.

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Rad21, also called SCC1 (sister chromatid cohesion protein 1), is part of the mitotic cohesin complex that consist of SMC1, SMC3, SCC3 and SCC1. The cohesin complex is important to sister chromatid separation and segregation during mitosis and meiosis and also plays a role in double-strand break repair by homologous recombination. Rad21 has been demonstrated to be cleaved by caspase proteins to trigger sister chromatid separation and during apoptotic signaling.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Comparar elementos similares

Ver comparación completa

Mostrar Diferencias

1 of 1

Este artículo
SAB4502562HPA020044HPA053282
clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

species reactivity

human, mouse

species reactivity

mouse, human

species reactivity

human

species reactivity

human

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Pruebe nuestra Herramienta de selección de productos para limitar sus opciones


Clase de almacenamiento

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Elija entre una de las versiones más recientes:

Certificados de análisis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documentos section.

Si necesita más asistencia, póngase en contacto con Atención al cliente

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos







Número de artículo de comercio global

SKUGTIN
PLA0022-100UL04061837721540

Questions

Reviews

No rating value

Active Filters