Saltar a
I8014
Inborn Errors of Metabolism, 21st ed.
Poster, 22 × 27 in.
Iniciar sesión para ver los precios por organización y contrato.
Seleccione un Tamaño
Cambiar Vistas
A ustedes/SKUDisponibilidadPrecio
Acerca de este artículo
NACRES:
NA.28
UNSPSC Code:
60104506
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarle
publication info
D.E. Nicholson, Sigma Chemical Co., St. Louis, MO, 1998
General description
Every characteristic of human anatomy and physiology is determined by biochemical reactions catalyzed by enzymes. These in turn are determined by our genetic make-up. If a gene is defective or missing it will result in a defective or missing enzyme, a so-called "inborn error of metabolism." Knowledge of the defective enzyme and its gene(s) may well make possible the rational treatment of the resulting disease. The Inborn Errors of Metabolism Map identifies the site of over 100 metabolic deficiency diseases which are named on the sides of the map.
Comparar elementos similares
Ver comparación completa
Mostrar Diferencias
1 of 1
Este artículo
NHPMHMAG-45K
publication info
D.E. Nicholson, Sigma Chemical Co., St. Louis, MO, 1998
publication info
-
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Questions
  1. Do you make a large poster for a wall? Say approximately 8-10 feet tall?
    1 answer
    1. Unfortunately, the 22 x 27 in. size is the only option available. Should a custom quote be of interest, please navigate to the link https://www.sigmaaldrich.com/support/customer-support, and click on "Quotes" under the Orders Section with all the required information so that a Sales team member can assist.