Seleccione un Tamaño
| Tamaño de envase | SKU | Disponibilidad | Precio |
|---|---|---|---|
| 100 μL | Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde) | $667.00 |
Acerca de este artículo
$667.00
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
product line
Prestige Antibodies® Powered by Atlas Antibodies
form
buffered aqueous glycerol solution
species reactivity
human
enhanced validation
orthogonal RNAseq
Learn more about Antibody Enhanced Validation
technique(s)
immunohistochemistry: 1:200-1:500
immunogen sequence
PKKVKVDCDSFLCQNPGEPGLQEAQKAGGLPADASPLFRQLFLKSQEPLVSHEQMQVFQMITKSQRIFSHAQV
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... ZNF541(84215)
Immunogen
Sequence
PKKVKVDCDSFLCQNPGEPGLQEAQKAGGLPADASPLFRQLFLKSQEPLVSHEQMQVFQMITKSQRIFSHAQV
Application
Features and Benefits
Every Prestige Antibody is tested in the following ways:
- IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
- Protein array of 364 human recombinant protein fragments.
Physical form
Other Notes
Legal Information
Disclaimer
1 of 1
Este artículo | |||
|---|---|---|---|
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| product line Prestige Antibodies® Powered by Atlas Antibodies | product line Prestige Antibodies® Powered by Atlas Antibodies | product line Prestige Antibodies® Powered by Atlas Antibodies | product line Prestige Antibodies® Powered by Atlas Antibodies |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
| Gene Information human ... ZNF541(84215) | Gene Information human ... ZNF541(84215) | Gene Information human ... ZNF544(27300) | Gene Information human ... ZNF514(84874) |
Still not finding the right product?
Pruebe nuestra Herramienta de selección de productos para limitar sus opciones
flash_point_f
Not applicable
flash_point_c
Not applicable
Clase de almacenamiento
10 - Combustible liquids
wgk
WGK 1
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?
Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.
Número de artículo de comercio global
| SKU | GTIN |
|---|---|
| HPA076453-100UL | 04061835313297 |
| HPA076453-25UL | 04061841146612 |



