Saltar a
T4949
Taprostene sodium salt
≥98% (HPLC)
Sinónimos:
Rheocyclan, Sodium 3-[(Z)-[(3aR,4R,5R,6aS)-4-[(1E,3S)-3-cyclohexyl-3-hydroxy-1-propenyl]hexahydro-5-hydroxy-2H-cyclopenta[b]furan-2-ylidene]methyl]benzoate
Iniciar sesión para ver los precios por organización y contrato.
Acerca de este artículo
Fórmula empírica (notación de Hill):
C24H29NaO5
Número CAS:
Peso molecular:
420.47
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Assay:
≥98% (HPLC)
assay
≥98% (HPLC)
color
off-white
mp
173-182.5 °C (lit.)
solubility
H2O: 26 mg/mL
storage temp.
2-8°C
SMILES string
[Na+].[H][C@]12C[C@@H](O)[C@H](\C=C\[C@@H](O)C3CCCCC3)[C@@]1([H])C\C(O2)=C\c4cccc(c4)C([O-])=O
InChI
1S/C24H30O5.Na/c25-21(16-6-2-1-3-7-16)10-9-19-20-13-18(29-23(20)14-22(19)26)12-15-5-4-8-17(11-15)24(27)28;/h4-5,8-12,16,19-23,25-26H,1-3,6-7,13-14H2,(H,27,28);/q;+1/p-1/b10-9+,18-12-;/t19-,20-,21-,22-,23+;/m1./s1
InChI key
KPQVOJYDUCZQEQ-REHYUDDHSA-M
Application
Taprostene has been used as an IP receptor agonist and was found to increase cAMP expression in human bronchial epithelial cells1.
Biochem/physiol Actions
Highly selective IP1 prostanoid receptor agonist.
Preparation Note
Taprostene is soluble in water at 26 mg/ml.
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Clase de almacenamiento
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
M Heinz et al.
VASA. Zeitschrift fur Gefasskrankheiten, 25(1), 65-72 (1996-01-01)
In a double blind pilot study, we examined the effects of the stable prostacyclin derivate taprostene compared to a combination of aspirin and dipyridamole on platelet uptake and clinical outcome after peripheral percutaneous angioplasty. Taprostene was administered to 19 patients
A M Lefer et al.
Methods and findings in experimental and clinical pharmacology, 16(9), 623-631 (1994-11-01)
We examined the effects of taprostene (2.5 x 10(-8) M to 1 x 10(-7) M), a stable prostacyclin analog, on PMN-endothelial interaction (i.e., adherence) and subsequent vasocontraction and endothelial dysfunction. Taprostene effectively inhibited the adherence of leukotriene B4-stimulated autologous cat
O Michel et al.
ORL; journal for oto-rhino-laryngology and its related specialties, 56(6), 305-309 (1994-11-01)
A probable relation between prostaglandin formation and the pathophysiology of hearing loss led to the application of the potent vasodilator prostacyclin in the treatment of inner ear disorders. Two drugs may be administered to humans: (1) unstable natural prostacyclin (epoprostenol)
Número de artículo de comercio global
SKUGTIN
T4949-1MG04061826677179
T4949-5MG04061832787343