Saltar al contenido
Merck

Saltar a

MABF591

Anti-CD45.1 Antibody (mouse), PE-Cy7, clone A20

clone A20, 0.2 mg/mL, from mouse

Sinónimos:

Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen, L-CA, Lymphocyte antigen 5, Ly-5, T200, CD45

Iniciar sesión para ver los precios por organización y contrato.

Seleccione un Tamaño

Cambiar Vistas
A ustedes/SKUDisponibilidadPrecio
25 μg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
157,00 €

Acerca de este artículo

UNSPSC Code:
12352203
NACRES:
NA.44
eCl@ss:
32160702
Conjugate:
PE-Cy7
Clone:
A20, monoclonal
Application:
FACS, IHC, IP
Citations:
-

157,00 €


Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad

Solicite una orden a granel
Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarle


biological source

mouse

Quality Segment

conjugate

PE-Cy7

antibody form

purified antibody

antibody product type

primary antibodies

clone

A20, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

concentration

0.2 mg/mL

technique(s)

flow cytometry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ptprc(19264)

General description

The A20 antibody reacts with mouse CD45.1, also known as Ly5.1, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors. The A20 antibody is typically used as a leukocyte marker in Ly5.1 mouse strains: SJL/J, DA, STS/A and RIII. The antibody has been demonstrated to specific for CD45.1 and is not cross-reactive with CD45.2 bearing cells.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
This mouse monoclonal antibody Anti-CD45.1 (mouse), PE-Cy7, clone A20 validated for use in IP, IHC & Flow Cytometry for the detection of CD45.1.

Physical form

Purified Mouse Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Size Exclusion Chromatography

Preparation Note

It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Analysis Note

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 1 µg of this antibody detected CD45.1 in one million SJL splenocytes.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Comparar elementos similares

Ver comparación completa

Mostrar Diferencias

1 of 1

Este artículo
MABF590MABF430MABF428
clone

A20, monoclonal

clone

A20, monoclonal

clone

A20, monoclonal

clone

A20, monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

Gene Information

mouse ... Ptprc(19264)

Gene Information

mouse ... Ptprc(19264)

Gene Information

mouse ... Ptprc(19264)

Gene Information

mouse ... Ptprc(19264)

conjugate

PE-Cy7

conjugate

APC-Cy7

conjugate

PE

conjugate

Allophycocyanin conjugate

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody


Still not finding the right product?

Pruebe nuestra Herramienta de selección de productos para limitar sus opciones


Clase de almacenamiento

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificados de análisis (COA)

Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos



Preguntas

Revisiones

Sin puntuación

Filtros activos