Saltar a
263141
Neodymium
chips, 99.9% trace rare earth metals basis
Iniciar sesión para ver los precios por organización y contrato.
Acerca de este artículo
Fórmula empírica (notación de Hill):
Nd
Número CAS:
Peso molecular:
144.24
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141609
EC Number:
231-109-3
MDL number:
assay
99.9% trace rare earth metals basis
form
chips
reaction suitability
reagent type: catalyst
core: neodymium
resistivity
64.0 μΩ-cm, 20°C
bp
3074 °C (lit.)
mp
1021 °C (lit.)
density
7.003 g/mL at 25 °C (lit.)
SMILES string
[Nd]
InChI
1S/Nd
InChI key
QEFYFXOXNSNQGX-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Clase de almacenamiento
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Artículos
Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.
Guillaume Soulet et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(17), 6682-6687 (2013-04-10)
Continental ice sheets are a key component of the Earth's climate system, but their internal dynamics need to be further studied. Since the last deglaciation, the northern Eurasian Fennoscandian Ice Sheet (FIS) has been connected to the Black Sea (BS)
Angie Phelan et al.
Australian orthodontic journal, 28(2), 159-169 (2013-01-12)
The demand for clear sequential aligner therapy has increased dramatically in recent years. An improved system utilising small neodymium-iron-boron (NdFeB) magnetic attachments has been proposed to enhance appliance capabilities. The aim of the investigation was to analyse the force system
Ionut Mihalcea et al.
Inorganic chemistry, 51(18), 9610-9618 (2012-08-31)
Three series of mixed uranyl-lanthanide (Ce or Nd) carboxylate coordination polymers have been successfully synthesized by means of a hydrothermal route using either conventional or microwave heating methods. These compounds have been prepared from mixtures of uranyl nitrate, lanthanide nitrate
Número de artículo de comercio global
SKUGTIN
263141-25G04061826117071