Skip to Content
Merck

Skip To

52411

Heptafluorobutyric acid

suitable for ion chromatography, ≥99.5% (GC)

Synonym(s):

Edman Reagent No. 3, HFBA, Perfluorobutyric acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CF3CF2CF2COOH
CAS Number:
Molecular Weight:
214.04
UNSPSC Code:
41115710
NACRES:
NB.21
PubChem Substance ID:
EC Number:
206-786-3
Beilstein/REAXYS Number:
1426882
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

7 (vs air)

Quality Level

vapor pressure

~10 mmHg ( 25 °C)

description

anionic

assay

≥99.5% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

technique(s)

ion chromatography: suitable

refractive index

n20/D 1.3 (lit.)

bp

120 °C/755 mmHg (lit.)

density

1.645 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg, Ba: ≤0.01 mg/kg, Bi: ≤0.01 mg/kg, Ca: ≤2 mg/kg, Cd: ≤0.01 mg/kg, Co: ≤0.01 mg/kg, Cr: ≤0.01 mg/kg, Cu: ≤0.01 mg/kg, Fe: ≤0.1 mg/kg, K: ≤0.5 mg/kg, Li: ≤0.01 mg/kg, Mg: ≤1 mg/kg, Mn: ≤0.01 mg/kg, Mo: ≤0.01 mg/kg, Na: ≤3 mg/kg, Ni: ≤0.01 mg/kg, Pb: ≤0.01 mg/kg, Sr: ≤0.01 mg/kg, Zn: ≤0.05 mg/kg

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C4HF7O2/c5-2(6,1(12)13)3(7,8)4(9,10)11/h(H,12,13)

InChI key

YPJUNDFVDDCYIH-UHFFFAOYSA-N

Application

Heptafluorobutyric acid (HFBA) may be used to cleave glycylserine bonds, in an experimental protocol done in order to sequence glycine-rich protein from lizard claw.

Other Notes

Visit the IC Portal to learn more

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
7724949540H1006
assay

≥99.5% (GC)

assay

≥99.0% (GC)

assay

-

assay

≥98%

technique(s)

ion chromatography: suitable

technique(s)

-

technique(s)

ion pair chromatography: suitable

technique(s)

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

liquid

form

liquid

form

liquid

form

liquid

density

1.645 g/mL at 25 °C (lit.)

density

1.645 g/mL at 25 °C (lit.)

density

-

density

1.674 g/mL at 20 °C (lit.)

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-


Still not finding the right product?

Explore all of our products under Heptafluorobutyric acid


signalword

Danger

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter

pictograms

CorrosionExclamation mark

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sequence of a glycine-rich protein from lizard claw: unusual dilute acid and heptafluorobutyric acid cleavages.
Inglis A
Proteins: Structure, Function, and Genetics, 757-764 (1987)
Ulrika Eriksson et al.
Environmental science and pollution research international, 20(11), 7940-7948 (2013-04-17)
Diet and drinking water are suggested to be major exposure pathways for perfluoroalkyl substances (PFASs). In this study, food items and water from Faroe Islands sampled in 2011/2012 were analyzed for 11 perfluoroalkyl carboxylic acids (PFCAs) and 4 perfluoroalkane sulfonic
Jan Busch et al.
Environmental pollution (Barking, Essex : 1987), 158(5), 1467-1471 (2010-01-08)
Polyfluoroalkyl compounds (PFCs) are widely used in industry and consumer products. These products could end up finally in landfills where their leachates are a potential source for PFCs into the aqueous environment. In this study, samples of untreated and treated



Global Trade Item Number

SKUGTIN
52411-25ML-F04061838261014
52411-INTR-F04061826678886
52411-5ML-F04061832549538

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters