Skip to Content
Merck
All Photos(1)

Key Documents

86854

Supelco

Tetrabutylammonium hydroxide solution

~40% in water, suitable for ion chromatography

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OH)
CAS Number:
Molecular Weight:
259.47
Beilstein:
3571228
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21
Pricing and availability is not currently available.

description

cationic

Quality Level

form

liquid

concentration

~40% in water

technique(s)

ion chromatography: suitable

impurities

≤0.01% halides (as bromide)

density

0.995 g/cm3

anion traces

sulfate (SO42-): ≤100 mg/kg

cation traces

K: ≤100 mg/kg
Na: ≤100 mg/kg

SMILES string

[OH-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H2/q+1;/p-1

InChI key

VDZOOKBUILJEDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium hydroxide (TBAOH) is a quaternary ammonium salt,mainly used as a strong base and phase-transfer catalyst.

Application

TBAOH may be used as an ion-pairing reagent in mobile phase preparation for reverse phase ion-pair high performance liquid chromatography during chromium (III) and chromium (VI) determination.[1]

Caution

may crystallize; can be redissolved on heating (max. 40°C)

Linkage

Visit the IC Portal to learn more

Other Notes

Reagent suitable for use in ion chromatographycitationcitation
Reagent suitable for use in ion chromatography[2][3]

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P.R. Haddad et al.
Ion Chromatography: Principles and Applications null
Simultaneous determination of chromium (III) and chromium (VI) with reversed-phase ion-pair high-performance liquid chromatography.
Jen, Jen-Fon, et al.
Analyst, 118.10 , 1281-1284 (1993)
T. Tang et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 45, 58-58 (1992)
Liwen Zhang et al.
ACS nano, 8(12), 12250-12258 (2014-11-18)
Photothermal therapy (PTT) is an emerging treatment modality that is under intensive preclinical investigations for the treatment of various medical conditions, including cancer. However, the lack of targeting function of PTT agents hampers its clinical application. An effective and nontoxic
Fabio Galeotti et al.
Journal of chromatography. A, 1284, 141-147 (2013-03-05)
In this study, by using tetrabutylammonium bisulfate as ion-pairing reagent, we were able to separate all the main heparin/heparan sulfate disaccharides generated by the action of heparinases along with the main Hep tetrasaccharide possessing a 3-O-sulfate group on the sulfoglucosamine

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service