Skip to Content
Merck
All Photos(2)

Key Documents

00612

Supelco

Water

suitable for ion chromatography

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12191602
PubChem Substance ID:
NACRES:
NB.21

Quality Level

technique(s)

ion chromatography: suitable

impurities

≤10 μg/kg acetate
≤10 μg/kg formiate
≤10 μg/kg glycolate
≤10 μg/kg oxalate

refractive index

n20/D 1.34 (lit.)

conductivity

≤2 μS/cm

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

anion traces

bromate (BrO3-): ≤1 μg/kg
bromide (Br-): ≤1 μg/kg
chlorate (ClO3-): ≤1 μg/kg
chloride (Cl-): ≤1 μg/kg
chlorite (ClO2-): ≤1 μg/kg
fluoride (F-): ≤1 μg/kg
iodide (I-): ≤1 μg/kg
nitrate (NO3-): ≤1 μg/kg
nitrite (NO2-): ≤1 μg/kg
phosphate (PO43-): ≤1 μg/kg
sulfate (SO42-): ≤1 μg/kg

cation traces

Al: ≤0.005 mg/kg
Ba: ≤0.005 mg/kg
Bi: ≤0.005 mg/kg
Ca: ≤0.01 mg/kg
Cd: ≤0.005 mg/kg
Co: ≤0.005 mg/kg
Cr: ≤0.005 mg/kg
Cu: ≤0.005 mg/kg
Fe: ≤0.005 mg/kg
K: ≤0.01 mg/kg
Li: ≤0.005 mg/kg
Mg: ≤0.005 mg/kg
Mn: ≤0.005 mg/kg
Mo: ≤0.005 mg/kg
NH4+: ≤0.01 mg/kg
Na: ≤0.01 mg/kg
Ni: ≤0.005 mg/kg
Pb: ≤0.005 mg/kg
Sr: ≤0.005 mg/kg
Zn: ≤0.005 mg/kg

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Water is of high purity and is used especially for the needs of ion chromatography (IC). It is suitable for trace analysis of anions, cations and some organics, which are usually analyzed by ion chromatography (IC).

Application

Metrohm IC application note AN-S-336:Anion traces on the Metrosep A Supp 16 - 250/2.0 after Inline Preconcentration and Matrix Elimination

Metrohm IC application note AN-Q-008:Trace monitoring in distilled water using ion chromatography.

Metrohm IC application note AN-C-159:Cation traces on the Metrosep C 4 - 250/2.0 column after Inline Preconcentration with matrix elimination.

Metrohm IC application note AN-S-304:Variable Inline Preconcentration including Matrix Elimination for trace anion analysis (MiPCT-ME).

Linkage

Visit the IC Portal to learn more

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Lieveld et al.
Virchows Archiv : an international journal of pathology, 465(6), 703-713 (2014-10-12)
Giant cell tumor of bone (GCTB) displays worrisome clinical features such as local recurrence and occasionally metastatic disease which are unpredictable by morphology. Additional routinely usable biomarkers do not exist. Gene expression profiles of six clinically defined groups of GCTB
Rachele M Pojednic et al.
Calcified tissue international, 96(3), 256-263 (2014-12-07)
Vitamin D receptor (VDR) expression and action in non-human skeletal muscle have recently been reported in several studies, yet data on the activity and expression of VDR in human muscle cells are scarce. We conducted a series of studies to
Boris Nowka et al.
Applied and environmental microbiology, 81(2), 745-753 (2014-11-16)
Nitrification has an immense impact on nitrogen cycling in natural ecosystems and in wastewater treatment plants. Mathematical models function as tools to capture the complexity of these biological systems, but kinetic parameters especially of nitrite-oxidizing bacteria (NOB) are lacking because
Mary A Rodgers et al.
The Journal of experimental medicine, 211(7), 1333-1347 (2014-06-25)
Linear ubiquitination is a newly discovered posttranslational modification that is currently restricted to a small number of known protein substrates. The linear ubiquitination assembly complex (LUBAC), consisting of HOIL-1L, HOIP, and Sharpin, has been reported to activate NF-κB-mediated transcription in
Liming Zhou et al.
Nature, 509(7498), 86-90 (2014-04-25)
Tropical forests are global epicentres of biodiversity and important modulators of climate change, and are mainly constrained by rainfall patterns. The severe short-term droughts that occurred recently in Amazonia have drawn attention to the vulnerability of tropical forests to climatic

Articles

Explore the tests integral to the instrument qualification process for ion chromatography instruments. Learn about the effectiveness of a pre-packaged IQ/OQ/PQ kit, featuring six certified bromide standard solutions for comprehensive IC instrument qualification tests.

Learn more about Bioethanol as a renewable, alternative fuel traditionally produced by the yeast fermentation of sugar.

Learn more about Bioethanol as a renewable, alternative fuel traditionally produced by the yeast fermentation of sugar.

Learn more about Bioethanol as a renewable, alternative fuel traditionally produced by the yeast fermentation of sugar.

See All

Protocols

Separation of Methane; Acetylene; Carbon monoxide; Water; Nitrogen; Carbon dioxide; Ethane; Ethylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service