MilliporeSigma
Mouse Embryo Media
Mouse whole embryo culture is an important method employed in mammalian developmental biology and birth defects research. This technique enables longitudinal studies of explanted rodent embryos at the organogenesis stage. An essential technique for creating transgenic mice uses the transformation and injection of stably transfected mouse embryonic stem cells (ES) into mouse blastocysts. To optimize embryo collection, manipulation, and transfer techniques, we offer a complete selection of mouse embryo media and reagents.
Products
Sort by Relevance
Showing 1-20 of 40
Change View
Showing 1-20 of 40
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 40 results
Page 1 of 2
Page 1 of 2
Hepes-based media for embryo handling and manipulation
  • M2 media is often used for embryo collection, and for extended handling outside the culture incubator
  • FHM is a modification of the KSOM formulation, also used for manipulating embryos outside CO2 incubation environments
Bicarbonate-based media for the culture and development of embryos
  • Modified M16 media: traditional embryo culture media based on Krebs-Ringer bicarbonate buffer
  • KSOM media allows embryos to overcome the two-cell block and supports enhanced cell division.  Our proprietary formulation provides higher yields of blastocyst development
All media are tested in mouse embryo culture and manufactured using premium quality raw materials.
EmbryoMax® Advanced KSOM Medium
EmbryoMax® Advanced KSOM Medium is an all-in-one medium for the harvest and culture of mouse embryos, replacing the traditional two-step system where unique, separate formulations are required for embryo harvest and culture. Our advanced KSOM formulation enables both handling of embryos in atmospheric conditions and culturing in a CO2-controlled incubator - with a single medium.
Six Stages of Embryo Development
Illustration of two round cells without nuclei, enclosed in a blue circle, representing the zygote stage of embryonic development.
Day 0
Zygote
    Illustration of two round cells with blue nuclei, enclosed in a blue circle, representing the 2-cell stage of embryonic development.
    2-cell
      Illustration of four round cells with blue nuclei, enclosed in a blue circle, representing the 4-cell stage of embryonic development.
      4-cell
        Illustration of a group of round cells with hollow centers, enclosed in a blue circle.
        Multi-cell
          Illustration of a cluster of small, round cells enclosed in a blue circle, representing a morula.
          Morula
            Illustration of numerous small, round cells clustered at the bottom of a blue circle, representing a blastocyst hatching.
            Blastocyst