Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Heptafluorobutyric acid
Synonym(s):
Edman Reagent No. 3, HFBA, Perfluorobutyric acid, Heptafluorobutyric acid
Linear Formula:
CF
3
CF
2
CF
2
COOH
CAS Number:
375-22-4
Molecular Weight:
214.04
EC Number:
206-786-3
Products
Product Category
organic acids (2)
acids (1)
building blocks (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Analyte Suitability
Quality Segment
Functional Group
Markush Class
Markush Group
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
164194
Heptafluorobutyric acid,
98%
Expand
View Pricing
77249
Heptafluorobutyric acid,
≥99.0% (GC)
Expand
View Pricing
8.43443
Heptafluorobutyric acid,
for synthesis
Expand
View Pricing
52411
Heptafluorobutyric acid,
suitable for ion chromatography, ≥99.5% (GC)
Expand
View Pricing