61699
Potassium hydroxide concentrate
0.1 M KOH in water (0.1N), Eluent concentrate for IC
Synonym(s):
Potassium hydroxide solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38100303
PubChem Substance ID:
NACRES:
NB.21
Recommended Products
Quality Level
concentration
0.1 M KOH in water (0.1N)
technique(s)
ion chromatography: suitable
SMILES string
[K+].[O-][H]
InChI
1S/K.H2O/h;1H2/q+1;/p-1
InChI key
KWYUFKZDYYNOTN-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Potassium hydroxide (KOH) solution is a strong alkali widely used in chemical synthesis. Van Slyke apparatus and Hildebrand adsorption technique have been utilized to determine the solubility of oxygen in different concentration of KOH solution.
This eluent concentrate for ion chromatography is determined by potentiometric titration. Content and expiry date can be found on the certificate.
Application
Potassium hydroxide solution may be used in the following processes:
- Depolymerization of poly(ethylene terephthalate) (PET) flakes.
- Synthesis of potassium bicarbonate and potassium carbonate.
- As an eluent in the nonsuppressor ion chromatography to separate and determine inorganic and organic anions.
- Saponification of fruit seed oil to determine tocopherols and tocotrienols.
Linkage
Visit the IC Portal to learn more
Preparation Note
Prepared with potassium hydroxide and high purity water (18.2 MΩ, 0.2 μm filtered)
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Native oils from apple, blackcurrant, raspberry, and strawberry seeds as a source of polyenoic fatty acids, tocochromanols, and phytosterols: a health implication.
Pieszka M, et al.
Journal of Chemistry, 2015 null
The Preparatory Manual of Explosives., 79-79 (2007)
Nonsuppressor ion chromatography of inorganic and organic anions with potassium hydroxide as eluent.
Okada T and Kuwamoto T.
Analytical Chemistry, 55 (7), 1001-1004 (1983)
Kinetics of depolymerization of poly (ethylene terephthalate) in a potassium hydroxide solution.
Industrial & Engineering Chemistry Research, 40 (2), 509-514 (2001)
The solubility and diffusion coefficient of oxygen in potassium hydroxide solutions.
Davis RE, et al.
Electrochimica Acta, 12 (3), 287-289 (1967)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service