Filter Housings
We have the expertise in designing, engineering, and manufacturing housings that satisfies the needs of the biopharmaceutical industry, as well as the rigorous requirements of global standards organizations. Our housings offer safe, reliable performance and process separation consistency to meet the complex needs of a wide range of your process applications.
Read more about
Related Product Resources
- Series 4000™ Single-round Filter Housings
- Series 4000™ Filter Housings User Guide
- Aseptic Filter Formats: Selection Guide
This technical article provides a high level overview of different formats available to help guide product selection.
Our Series 4000™ single-round filter housing line provides a range of pre-configured designs to meet all your process and filtration needs. It is available in T-Line and In-Line designs, with a wide range of tube sizes.
- Available and ready to be shipped to your location
- Low dead volume to minimize product loss
- Enhanced cleanability (CIP/SIP)
- Enhanced documentation package, to ensure regulatory compliance
Proven stainless steel components are used in all housings, ensuring consistent quality and performance. Select sizes are available with a Code 7 base dedicated to critical applications, and a full range of cartridges to suit your application requirements.
Millidisk® Sanitary Cartridge Housing
Millidisk® cartridge housings are designed to handle the most critical sterile filtration steps for industrial processing applications. Typical applications include: sterile filtration of aqueous and solvent-based solutions, sterile tank venting, and sterile gas filtration.
Milligard® Sanitary Cartridge Housings
The Milligard® housing is ideal for use with Millipore Code M cartridges in liquid or gas.
Millistak+® and Millistak+® HC Housings
Millistak+® 316L stainless steel filter housings are designed for high-capacity liquid clarification. Offered in multiple sizes to accommodate varying production process volumes, Millistak+® housings complement the removal capacity, dependable performance, and efficiency of our extensive line of Millistak+® and Millistak+® HC filtration cartridges.
Optiseal® Sanitary Cartridge Housings
The Optiseal® housing is designed for sterile filtration of aqueous solutions and gases using Optiseal® cartridge filters. The housing features easy-to-clean vents, drains, and connections, allowing complete sterilization.