Skip to Content
Merck
All Photos(1)

Key Documents

50972

Supelco

2,6-Pyridinedicarboxylic acid concentrate

0.02 M C7H5NO4 in water (0.04N), suitable for ion chromatography, eluent concentrate

Synonym(s):

2,6-Pyridinedicarboxylic acid solution, Dipicolinic acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO4
CAS Number:
Molecular Weight:
167.12
MDL number:
UNSPSC Code:
12000000
eCl@ss:
39151816
PubChem Substance ID:
NACRES:
NB.21
Pricing and availability is not currently available.

description

anionic

Quality Level

concentration

0.02 M C7H5NO4 in water (0.04N)

technique(s)

ion chromatography: suitable

SMILES string

OC(=O)c1cccc(n1)C(O)=O

InChI

1S/C7H5NO4/c9-6(10)4-2-1-3-5(8-4)7(11)12/h1-3H,(H,9,10)(H,11,12)

InChI key

WJJMNDUMQPNECX-UHFFFAOYSA-N

General description

This eluent concentrate for ion chromatography is determined by potentiometric titration. Content and expiry date can be found on the certificate.

Application

Metrohm IC application note AN-C-163: Cations inbrine with minimal dilution and sub-μL injection.

Metrohm IC application note AN-C-172: Cations insnow from an open field.

Metrohm IC application note AN-C-143: Ammonia inaddition to standard cations in maritime pore water.

Linkage

Visit the IC Portal to learn more

Preparation Note

Prepared with 2,6-pyridinecarboxylic acid and high purity water (18.2 MΩ, 0.2 μm filtered)

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chao Xu et al.
Inorganic chemistry, 52(5), 2750-2756 (2013-02-21)
Complexation of UO2(2+) with dipicolinic acid (DPA) has been investigated in 0.1 M NaClO4. The stability constants (log β1 and log β2) for two successive complexes, UO2L and UO2L2(2-) where L(2-) stands for the deprotonated dipicolinate anion, were determined to
C C J van Melis et al.
International journal of food microbiology, 160(2), 124-130 (2012-11-28)
In this study, the impact of a range of organic acids and structurally similar alcohols with three to six carbon backbones and increasing lipophilic character, were tested on the germination behavior of B. cereus ATCC 14579 spores. This approach allowed
Pengfei Zhang et al.
Journal of bacteriology, 194(3), 646-652 (2011-11-30)
The kinetic parameters of the release of Ca(2+)-dipicolinic acid (CaDPA) during germination of spore populations and multiple individual spores of Bacillus subtilis strains with major alterations in the structure of the spore peptidoglycan (PG) cortex or lacking one or both
Xuan Yi Butzin et al.
Journal of bacteriology, 194(21), 5749-5758 (2012-08-21)
As previously reported, gerP Bacillus subtilis spores were defective in nutrient germination triggered via various germinant receptors (GRs), and the defect was eliminated by severe spore coat defects. The gerP spores' GR-dependent germination had a longer lag time between addition
Geneviève Labbé et al.
Journal of inorganic biochemistry, 112, 49-58 (2012-05-02)
It has long been suggested that the essential and ubiquitous enzyme fructose 1,6-bisphosphate (FBP) aldolase could be a good drug target against bacteria and fungi, since lower organisms possess a metal-dependant (Class II) FBP aldolase, as opposed to higher organisms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service