MilliporeSigma
Filter Aids and Filter Media
Sea sand
Filter aids, also known as filtration aids or filter media, are substances or materials widely used to enhance the efficiency of filtration processes. These finely graded materials are added to a liquid to be filtered, helping to remove impurities that may otherwise clog the filter medium. A filter aid functions as an agent composed of solid particles that improve filtering efficiency by creating a porous, permeable, and rigid lattice structure known as the filter cake. This filter cake retains solid particles and also contributes to flow control. Filtering aids find application in diverse industries such as pharmaceuticals, food and beverage, water treatment, and manufacturing.
Common types of filter aids include activated carbon, sand, diatomaceous earth, and cellulose. These substances enhance filtration processes in various industries by offering high porosity, adsorption capabilities, biodegradability, and efficient removal of impurities.
Section Overview
Activated Carbon Filters
Activated carbon is a commonly used filtering agent due to its excellent adsorption properties. It is a form of carbon subjected to special processing to generate a highly porous structure with a substantial surface area. This porous structure allows activated carbon to effectively trap and adsorb impurities from gases, liquids, or vapors. Applications of activated carbon include adsorbing organic compounds, colorants, odors, and specific chemicals from liquids or gases.
Sea Sand Filters
Sea sand, composed primarily of silicon oxide with trace amounts of other metal oxides, is a natural product known for its versatility and efficiency in scientific applications. It is widely used in laboratories for the filtration and clarification of heavily contaminated water and solutions. Additionally, sea sand serves as a grinding aid and is employed in heating baths (sand baths). To meet the high-quality standards required for laboratory use, sea sand is first boiled in hydrochloric acid, then ignited to carbonize and remove organic contaminants.  This process effectively eliminates acid-soluble impurities, making sea sand suitable for a range of scientific purposes.  
Diatomaceous Earth (DE) Filters
Diatomaceous earth is a naturally occurring amorphous silica formed from the fossilized skeletons of diatoms. Diatomaceous earth filter aid is widely used due to its high water and oil absorption abilities. Its pronounced porosity makes it highly effective at trapping fine particles. Additionally, it can serve as a silica precursor for adsorbing various oil groups, including benzene, toluene, ethylbenzenes, p-xylenes, and o-xylenes.
Cellulose Filters
Cellulose can be used as a filtering agent due to its porous and absorbent nature. The fibrous structure of cellulose forms an interconnected network of channels, facilitating fluid passage while capturing particles and impurities. Cellulose-based filters are commonly used in various industries, including water treatment, pharmaceutical processing, food and beverage production, and laboratory applications. These filters effectively separate solids from liquids and can help remove contaminants, such as sediment, bacteria, and other particulates, ensuring cleaner and purified fluids. Moreover, cellulose powder and regenerated cellulose are used in chromatography processes.
Related Product Resources