Skip to Content
MilliporeSigma

Skip To

PFS22

SpheriCal Neat

Peptide High, suitable for MALDI-MS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

SpheriCal

form

solid

shelf life

expiry date on the label

mol wt

Mr 3500-7500 Da (Na+-adduct)

technique(s)

MALDI-MS: suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

−20°C

General description

SpheriCal is a dendrimer-based MALDI calibration kit providing 4 monodisperse mass signals per kit for efficient, highly accurate calibration. By combining excellent long-term stability with evenly-spaced calibration points, SpheriCal helps users increase the frequency and quality of their MALDI-MS calibrations. With a shelf life of years and each kit offering at least 100 calibrations, SpheriCal offers a reliable and accurate calibration across a large mass range, without the shelf-life and storage issues associated with protein or peptide-based systems.

Mass ranges available:

i.SpheriCal Neat Peptide Low (m/z 500 - 1,600)
ii.SpheriCal Neat Peptide Medium (m/z 1,600 - 3,500)
iii.SpheriCal Neat Peptide High (m/z 3,500 - 7,500)
iv.SpheriCal Neat Protein Low (m/z 7,500 - 15,000)
v.SpheriCal Neat Protein Medium(m/z 15,000 - 30,000).

SpheriCal Neat: unprecedented freedom for the user.

SpheriCal Neat is provided without any matrix or added cation and, because the SpheriCal calibration standards exhibit unusually broad solvent and matrix compatibility, this product can be used with a wide range of sample preparation conditions, though ionization is preferred with alkali cations. The freedom of this product line will enable you to explore your own calibration protocols for optimization purposes. Each vial of SpheriCal Neat contains sufficient calibration standard for over 100 calibration spots.

Find more information here SpheriCal
dendritic calibrants PFS3A : PFS3B : PFS3C : PFS3D = 1:1:1:1

Application

For mass calibration in mass spectrometry

Packaging

Quantity: 50 μg/vial

Legal Information

SpheriCal is a trademark of Polymer Factory Sweden AB


Still not finding the right product?

Explore all of our products under SpheriCal Neat


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

SpheriCal MALDI Calibration Guide Neat and Mix





Global Trade Item Number

SKUGTIN
PFS22-1EA04061834398592

Questions

Reviews

No rating value

Active Filters