Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56229

Supelco

9-Nitroanthracene

matrix substance for MALDI-MS, ≥98.5% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9NO2
CAS Number:
Molecular Weight:
223.23
Beilstein:
1877509
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥98.5% (HPLC)

technique(s)

MALDI-MS: suitable

mp

141-144 °C (lit.)
144-150 °C

cation traces

Ba: ≤5 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤100 mg/kg
K: ≤20 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

suitability

suitable for matrix substance for MALDI-MS

SMILES string

[O-][N+](=O)c1c2ccccc2cc3ccccc13

InChI

1S/C14H9NO2/c16-15(17)14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-9H

InChI key

LSIKFJXEYJIZNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service