Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83788

Supelco

(2E)-3-(9-Anthryl)-2-cyanoacrylic acid

matrix substance for MALDI-MS, ≥97.0% (HPLC)

Synonym(s):

(2E)-3-(Anthracen-9-yl)-2-cyanoacrylic acid, (2E)-3-(Anthracen-9-yl)-2-cyanoprop-2-enoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11NO2
Molecular Weight:
273.29
Beilstein:
21824385
UNSPSC Code:
12000000
NACRES:
NA.21

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥97.0% (HPLC)

form

powder

technique(s)

MALDI-MS: suitable

cation traces

Al: ≤20 mg/kg
Ba: ≤20 mg/kg
Ca: ≤20 mg/kg
Cd: ≤20 mg/kg
Co: ≤20 mg/kg
Cr: ≤20 mg/kg
Cu: ≤20 mg/kg
Fe: ≤20 mg/kg
K: ≤20 mg/kg
Li: ≤20 mg/kg
Mg: ≤20 mg/kg
Mn: ≤20 mg/kg
Mo: ≤20 mg/kg
Na: ≤20 mg/kg
Ni: ≤20 mg/kg
Pb: ≤20 mg/kg
Sr: ≤20 mg/kg
Zn: ≤20 mg/kg

suitability

suitable for matrix substance for MALDI-MS

Application

suitable as Maldi-Matrix material

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service