Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

02241

Supelco

2′,3′-Dideoxycytidine 5′-triphosphate sodium salt solution

suitable for MALDI-MS, ≥98.0% (HPLC)

Synonym(s):

ddCTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N3NaO12P3
CAS Number:
Molecular Weight:
473.14
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

biological source

animal

Quality Level

Assay

≥98.0% (HPLC)

form

liquid

packaging

pkg of 1 μmol

concentration

10.0-11.0 (UV)

technique(s)

MALDI-MS: suitable
mass spectrometry (MS): suitable

impurities

≤1.5% ddNDP (HPLC)

suitability

corresponds for UV test

storage temp.

−20°C

SMILES string

[Na+].NC1=NC(=O)N(C=C1)[C@H]2CC[C@@H](COP([O-])(=O)OP(O)(=O)OP(O)(O)=O)O2

InChI

1S/C9H16N3O12P3.Na/c10-7-3-4-12(9(13)11-7)8-2-1-6(22-8)5-21-26(17,18)24-27(19,20)23-25(14,15)16;/h3-4,6,8H,1-2,5H2,(H,17,18)(H,19,20)(H2,10,11,13)(H2,14,15,16);/q;+1/p-1/t6-,8+;/m0./s1

InChI key

LOOMQRBEBYCFPP-QDOHZIMISA-M

Biochem/physiol Actions

Inhibitor of DNA polymerase I-catalyzed chain elongation.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaligara Veeranagouda et al.
BMC genomics, 15, 429-429 (2014-06-06)
Bacteroides fragilis is a Gram-negative anaerobe that is normally a human gut commensal; it comprises a small percentage of the gut Bacteroides but is the most frequently isolated Bacteroides from human infections. Identification of the essential genes necessary for the
Nutjaree Jeenduang et al.
Clinica chimica acta; international journal of clinical chemistry, 442, 36-43 (2015-01-17)
Cardiovascular disease (CVD) is one of the major causes of death worldwide. Numerous genetic risk factors in lipid metabolism, including mutations of LDLR, APOB, and PCSK9, as well as polymorphisms of CETP and APOE, have been found to associate with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service