Skip to Content
Merck

Skip To

C8857

ProteoMass Cytochrome c MALDI-MS Standard

vial of 10 nmol, (M+H+) 12,361.96 Da by calculation

Synonym(s):

Cytochrome c from equine heart, Cytochrome c from horse heart

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

ProteoMass

Quality Segment

form

solid

mol wt

(M+H+) 12,361.96 Da by calculation

packaging

vial of 10 nmol

technique(s)

mass spectrometry (MS): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Application

Protein calibration standard for MALDI-MS instruments.
The specific sites and extent of oxidation in horse cytochrome c treated with H2O2 and UV were characterized. It was suggested that the state of these sites could be used as a biomarker for the oxidative environment in a cell.

Biochem/physiol Actions

Cytochrome c is primarily known as an electron-carrying mitochondrial protein. The transition of cytochrome c between the ferrous and ferric states within the cell makes it an efficient biological electron-transporter and it plays a vital role in cellular oxidations in both plants and animals. It is generally regarded as a universal catalyst of respiration, forming an essential electron-bridge between the respirable substrates and oxygen

Other Notes

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C2436C3131C2037
technique(s)

mass spectrometry (MS): suitable

technique(s)

cell based assay: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell based assay: suitable

product line

ProteoMass

product line

-

product line

-

product line

-

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cell analysis

application(s)

-

application(s)

cell analysis

mol wt

(M+H+) 12,361.96 Da by calculation

mol wt

12,588 Da

mol wt

12327 Da

mol wt

12327 Da

packaging

vial of 10 nmol

packaging

-

packaging

-

packaging

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

300


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Esther Lapuente-Brun et al.
Science (New York, N.Y.), 340(6140), 1567-1570 (2013-07-03)
The textbook description of mitochondrial respiratory complexes (RCs) views them as free-moving entities linked by the mobile carriers coenzyme Q (CoQ) and cytochrome c (cyt c). This model (known as the fluid model) is challenged by the proposal that all
Rawabi Qadhi et al.
Toxicology letters, 219(3), 288-297 (2013-03-26)
The importance of dietary polyunsaturated fatty acids (PUFAs) in the reduction of cardiovascular disease has been recognized for many years. Docosahexaenoic acid (22:6n3, DHA) is an n-3 PUFA known to affect numerous biological functions and provide cardioprotection; however, the exact
Keren I Hilgendorf et al.
Genes & development, 27(9), 1003-1015 (2013-04-27)
The retinoblastoma protein gene RB-1 is mutated in one-third of human tumors. Its protein product, pRB (retinoblastoma protein), functions as a transcriptional coregulator in many fundamental cellular processes. Here, we report a nonnuclear role for pRB in apoptosis induction via



Global Trade Item Number

SKUGTIN
C8857-1VL04061833296011
C8857-5X1VL04061838188076

Questions

Reviews

No rating value

Active Filters