Skip to Content
Merck
All Photos(1)

Documents

56451

Supelco

1,5-Diaminonaphthalene

matrix substance for MALDI-MS, ≥99.0% (HPLC)

Synonym(s):

1,5-DAN, 1,5-Naphthalenediamine, NSC 401110

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(NH2)2
CAS Number:
Molecular Weight:
158.20
Beilstein:
907947
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.0% (HPLC)

composition

carbon content, 75.92% (theory)
hydrogen content, 6.37% (theory)
nitrogen content, 17.71% (theory)

analyte chemical class(es)

oligonucleotides, peptides

technique(s)

MALDI-MS: suitable

mp

185-187 °C (lit.)

cation traces

Ba: ≤5.0 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5.0 mg/kg
Co: ≤5.0 mg/kg
Cr: ≤5.0 mg/kg
Cu: ≤5.0 mg/kg
K: ≤10 mg/kg
Mg: ≤5.0 mg/kg
Mn: ≤5.0 mg/kg
Na: ≤10 mg/kg
Ni: ≤5.0 mg/kg
Pb: ≤5.0 mg/kg
Zn: ≤5.0 mg/kg

SMILES string

Nc1cccc2c(N)cccc12

InChI

1S/C10H10N2/c11-9-5-1-3-7-8(9)4-2-6-10(7)12/h1-6H,11-12H2

InChI key

KQSABULTKYLFEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,5-Diaminonaphthalene is used in the production of 1,5-naphthalene diisocyanate and polyurethane elastomers.

Application

1,5-Diaminonaphthalene is used as an effective matrix in the analysis of IgG protein using top-down and middle-down mass spectrometry with multiple fragmentation techniques such as electron transfer dissociation (ETD) and matrix-assisted laser desorption ionization in-source decay (MALDI-ISD) method.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

metal trace analysis corresponds to requirements

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

438.8 °F - DIN 51758

Flash Point(C)

226 °C - DIN 51758


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delphine Debois et al.
Analytical chemistry, 82(10), 4036-4045 (2010-04-20)
Matrix-assisted laser desorption ionization-mass spectrometry (MALDI-MS) imaging is a powerful technique giving access to the distribution of a large range of biomolecules directly from a tissue section, allowing, for example, the discovery of new pathological biomarkers. Nevertheless, one main difficulty
Daiki Asakawa et al.
Journal of mass spectrometry : JMS, 47(2), 180-187 (2012-02-24)
Matrix-assisted laser desorption/ionization in-source decay (MALDI-ISD) has been used for characterization of a phosphorylated peptides and proteins because labile phosphate group is not lost during the MALDI-ISD process. The conventional MALDI-ISD is initiated by the hydrogen transfer from reducing matrix
Laura Molin et al.
Rapid communications in mass spectrometry : RCM, 25(20), 3091-3096 (2011-09-29)
1,5-Diaminonaphthalene (DAN) has been described as an interesting and effective matrix for matrix-assisted laser desorption/ionization (MALDI) experiments in positive ion mode, being able to activate in-source decomposition phenomena and, when employed for the analysis of proteins containing disulphide bridge(s), being
Aurélien Thomas et al.
Analytical chemistry, 84(4), 2048-2054 (2012-01-17)
Matrix sublimation has demonstrated to be a powerful approach for high-resolution matrix-assisted laser desorption ionization (MALDI) imaging of lipids, providing very homogeneous solvent-free deposition. This work presents a comprehensive study aiming to evaluate current and novel matrix candidates for high
Pascale Lemaire et al.
Rapid communications in mass spectrometry : RCM, 27(16), 1837-1846 (2013-07-17)
In-Source Decay (ISD) in Matrix-Assisted Laser Desorption/Ionization (MALDI) mass spectrometry is a fast and easy top-down activation method. Our objective is to find a suitable matrix to locate the deuterons following in-solution hydrogen/deuterium exchange (HDX). This matrix must circumvent the

Protocols

A short abstract on a method using QuEChERS and GC/MS/MS for determination of fipronil & fipronil sulfone.

A short abstract on a method using QuEChERS and GC/MS/MS for determination of fipronil & fipronil sulfone.

A short abstract on a method using QuEChERS and GC/MS/MS for determination of fipronil & fipronil sulfone.

A short abstract on a method using QuEChERS and GC/MS/MS for determination of fipronil & fipronil sulfone.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service