Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
bovine heart
Quality Segment
assay
≥95% based on Mol. Wt. 12,327 basis
form
powder or crystals
mol wt
12327 Da
technique(s)
cell based assay: suitable
suitability
suitable for molecular biology
UniProt accession no.
application(s)
cell analysis
storage temp.
−20°C
Gene Information
cow ... CYC1(512500)
General description
Application
- for cytochrome oxidase staining using the mice brain tissuePMID 27798184
- as a component of control mixture to evaluate the accuracy of tandem mass spectra in identifying peptidesPMID 12143966
- to study its effect on the stability, transport, and retention of biochar colloids (BCs) in saturated porous media under various solution pH and ionic strength (IS) conditionsPMID 30623842
- as a component of staining solution for the detection of cytochrome oxidase activity in the brain tissue sections of mousePMID 27798184
- as a component in the assay to study its superoxide scavenging efficiency
- as a component of serum-free DMEM media to evaluate non-specific cytochrome C reduction in astrocytes and HT-29 cellsPMID 25761243
Biochem/physiol Actions
Cytochrome c has been identified as an important mediator in apoptotic pathways. The release of mitochondrial cytochrome c into the cytoplasm stimulates apoptosis and is commonly used as an indicator of the apoptotic process in the cell. Investigation on the effect of Paris Saponin I (PS I) on human gastric carcinoma cell growth (SGC7901 cells) have shown an elevated level cytoplasmic cytochrome c. Results are an inhibition of proliferation in SGC7901 cells by inducing mitochondria-dependent apoptosis through cytochrome.
Preparation Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description ≥95% based on Mol. Wt. 12,327 basis | description ≥95% (SDS-PAGE) | description vial of 10 nmol, (M+H+) 12,361.96 Da by calculation | description BioReagent, suitable for GFC marker |
| assay ≥95% based on Mol. Wt. 12,327 basis | assay ≥95% (SDS-PAGE) | assay - | assay - |
| technique(s) cell based assay: suitable | technique(s) cell based assay: suitable | technique(s) mass spectrometry (MS): suitable | technique(s) - |
| biological source bovine heart | biological source horse heart | biological source - | biological source - |
| suitability suitable for molecular biology | suitability suitable for molecular biology | suitability - | suitability - |
| mol wt 12327 Da | mol wt 12,384 | mol wt (M+H+) 12,361.96 Da by calculation | mol wt ~12,400 |
| form powder or crystals | form powder | form solid | form powder |
Still not finding the right product?
Explore all of our products under Cytochrome c from bovine heart
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

