Skip to Content
Merck

Skip To

C3131

Cytochrome c from bovine heart

≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture

Synonym(s):

CYC, CytC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-700-9
NACRES:
NA.61
MDL number:
Form:
powder
Assay:
≥95% based on Mol. Wt. 12,327 basis
Biological source:
bovine heart
Mol wt:
12327 Da
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine heart

Quality Segment

assay

≥95% based on Mol. Wt. 12,327 basis

form

powder

mol wt

12327 Da

storage condition

(Tightly closed Dry)

technique(s)

cell culture | mammalian: suitable

impurities

0.40-0.50% Iron (anhydrous)

solubility

water: 10 mg/mL, dark red-brown

UniProt accession no.

storage temp.

−20°C

Gene Information

cow ... CYC1(512500)

General description

Research area: Apoptosis

Cytochrome c (Cyt C) is an essential mitochondrial protein located in the inner membrane of the mitochondria.[1] It is encoded in the nucleus as apo-cytochrome C.[2] Cyt C occurs as membrane-bound or as soluble metalloproteins in energy-transducing membranes and is found in bacteria, archaea, and plastids.[1] It belongs to the group of class IIa cytochromes and contains pentacoordinate heme centres.[3]

Application

Cytochrome C from bovine heart has been used:

  • as a component of the assay solution for testing the activity of respiratory chain complexes III and IV in the mitochondria of human osteosarcoma and human hepatocarcinoma cell lines[4]
  • to determine the cytochrome c oxidase activity in mitochondria of human aortic endothelial cells[5]
  • as a component of the cyt C oxidase reaction solution during in situ enzyme staining of cyt C oxidase in mice kidney tissues

Biochem/physiol Actions

Cytochrome C (Cyt C) is involved in the activation of caspase during the caspase-dependent apoptosis intrinsic pathway that triggers programmed cell death through apoptosis.[6] It shows lipid-binding activity through its interaction with cardiolipin that accounts for the peroxidase activity of Cyt C.[1] Cyt C binds to heme via the help of cytochrome c heme lyase that enables its release into the mitochondrial intermembrane space.[7]
The ready fluctuation of cytochrome c within the cell between ferrous and ferric states, makes it an efficient biological electron-transporter. It plays a vital role in cellular oxidations in both plants and animals. Generally regarded as a universal catalyst of respiration, it forms the essential electron-bridge between the respirable substrates and oxygen.
The ready interconversion of cytochrome c between ferrous and ferric states makes it an efficient biological electron carrier. It plays a vital role in cellular oxidations in both plants and animals. Generally regarded as a universal link in the respiratory chain, it forms the essential electron-bridge between the respirable substrates and oxygen.

Preparation Note

Prepared with acetic acid without using TCA.

Other Notes


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C2037C2867C7150
biological source

bovine heart

biological source

bovine heart

biological source

horse heart

biological source

-

assay

≥95% based on Mol. Wt. 12,327 basis

assay

≥95% based on Mol. Wt. 12,327 basis

assay

≥99% (SDS-PAGE)

assay

-

technique(s)

cell culture | mammalian: suitable

technique(s)

cell based assay: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

mol wt

12327 Da

mol wt

12327 Da

mol wt

12,384

mol wt

~12,400

form

powder

form

powder or crystals

form

powder

form

powder

Gene Information

cow ... CYC1(512500)

Gene Information

cow ... CYC1(512500)

Gene Information

horse ... CYCS(100053958)

Gene Information

-


Still not finding the right product?

Explore all of our products under Cytochrome c from bovine heart


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters