Skip to Content
Merck

Skip To

82170

(R)-Naproxen

pharmaceutical impurity standard

Synonym(s):

(R)-(−)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, Naproxen Impurity G, (2R)-2-(6-Methoxy-2-naphthyl)propionic acid, (2R)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (R)-Naproxen

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3OC10H6CH(CH3)CO2H
CAS Number:
Molecular Weight:
230.26
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
245-966-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

API family

naproxen

assay

≥98.0% (HPLC)

optical activity

[α]/D -71±4°, c = 0.1 in chloroform

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

mp

156-158 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc2cc(ccc2c1)[C@@H](C)C(O)=O

InChI

1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/t9-/m1/s1

InChI key

CMWTZPSULFXXJA-SECBINFHSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8082246482570842
application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

clinical
environmental
forensics and toxicology
pharmaceutical (small molecule)

application(s)

-

format

neat

format

neat

format

neat

format

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

assay

≥98.0% (HPLC)

assay

≥98.0% (HPLC)

assay

-

assay

98%

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

-


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
82170-50MG04061834246763

Questions

Reviews

No rating value

Active Filters