Skip to Content
Merck
All Photos(1)

Key Documents

46482

Supelco

Naproxen

VETRANAL®, analytical standard

Synonym(s):

(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CH(CH3)CO2H
CAS Number:
Molecular Weight:
230.26
Beilstein:
3591068
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

152-154 °C (lit.)

application(s)

clinical
environmental
forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

COc1ccc2cc(ccc2c1)[C@H](C)C(O)=O

InChI

1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/t9-/m0/s1

InChI key

CMWTZPSULFXXJA-VIFPVBQESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Haupt et al.
Journal of biochemical and biophysical methods, 25(4), 273-284 (1992-12-01)
A column-switching system for fast determination of (R)- and (S)-naproxen in liver microsomes has been developed. The centrifuged sample was injected directly onto a pre-column with octadecylcoated silica. The retained analytes were then directed to an alpha 1-AGP column using
Jiayang Li et al.
Journal of the American Chemical Society, 135(2), 542-545 (2012-11-10)
As systemically used therapeutics for treating acute or chronic pains or inflammations, nonsteroidal anti-inflammatory drugs (NSAIDs) also associate with the adverse gastrointestinal and renal effects and cardiovascular risks. Thus, it is beneficial to develop topical gels that selectively inhibit cyclooxygenase-2
Michela Campolo et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(11), 4489-4499 (2013-08-01)
Spinal cord trauma causes loss of motor function that is in part due to the ensuing inflammatory response. Hydrogen sulfide (H2S) is a potent, endogenous anti-inflammatory and neuroprotective substance that has been explored for use in the design of novel
J Rossat et al.
Clinical pharmacology and therapeutics, 66(1), 76-84 (1999-08-03)
To compare the renal hemodynamic and tubular effects of celecoxib, a selective inhibitor of cyclooxygenase-2 (COX-2) to those of naproxen, a nonselective inhibitor of cyclooxygenases in salt-depleted subjects. Forty subjects were randomized into four parallel groups to receive 200 mg
Dong Qing Zhang et al.
Chemosphere, 91(1), 14-21 (2012-12-27)
Scirpus validus was grown hydroponically and exposed to the pharmaceuticals, carbamazepine and naproxen at concentrations of 0.5-2.0 mg L(-1) for an exposure duration of up to 21 d. By the end of experiment, carbamazepine elimination from the nutrient solution reached

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service