Skip to Content
Merck

Skip To

07503

2-Bromo-6-methoxynaphthalene

pharmaceutical impurity standard

Synonym(s):

Naproxen Impurity N

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
BrC10H6OCH3
CAS Number:
Molecular Weight:
237.09
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
225-837-0
Beilstein/REAXYS Number:
2043874
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

API family

naproxen

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

mp

106-109 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1ccc2cc(Br)ccc2c1

InChI

1S/C11H9BrO/c1-13-11-5-3-8-6-10(12)4-2-9(8)7-11/h2-7H,1H3

InChI key

AYFJBMBVXWNYLT-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR33320754708137
format

neat

format

-

format

neat

format

neat

assay

≥98.0% (GC)

assay

-

assay

≥96.5% (GC)

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

impurities

-

impurities

≤0.5% water

impurities

≤0.5% water

Quality Level

100

Quality Level

300

Quality Level

100

Quality Level

100

mp

106-109 °C (lit.)

mp

-

mp

201-206 °C (lit.)

mp

-


Still not finding the right product?

Explore all of our products under 2-Bromo-6-methoxynaphthalene


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
199567-100G04061832338842
07503-50MG04061834242932

Questions

Reviews

No rating value

Active Filters