Skip to Content
MilliporeSigma

Skip To

1572503

USP

Propofol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2,6-Diisopropylphenol, 2,6-Bis(1-methylethyl)phenol, 2,6-Bis(isopropyl)phenol, Propofol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
[(CH3)2CH]2C6H3OH
CAS Number:
Molecular Weight:
178.27
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor pressure

5.6 mmHg ( 100 °C)

API family

propofol

manufacturer/tradename

USP

refractive index

n20/D 1.514 (lit.)

bp

256 °C/764 mmHg (lit.)

mp

18 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)c1cccc(C(C)C)c1O

InChI

1S/C12H18O/c1-8(2)10-6-5-7-11(9(3)4)12(10)13/h5-9,13H,1-4H3

InChI key

OLBCVFGFOZPWHH-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Propofol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Propofol Injectable Emulsion

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0000016BP1031PHR1663
Propofol British Pharmacopoeia (BP) Reference Standard

BP1031

Propofol

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

BP

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

density

0.962 g/mL at 25 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

Gene Information

human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

Gene Information

human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

Gene Information

human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

Gene Information

human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)


Still not finding the right product?

Explore all of our products under Propofol


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1572503-500MG04061835346264

Questions

Reviews

No rating value

Active Filters