Skip to Content
MilliporeSigma

Skip To

PHR1663

Propofol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2,6-Diisopropylphenol, 2,6-Bis(1-methylethyl)phenol, 2,6-Bis(isopropyl)phenol, Propofol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
[(CH3)2CH]2C6H3OH
CAS Number:
Molecular Weight:
178.27
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
218-206-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

grade

certified reference material, pharmaceutical secondary standard

agency

traceable to Ph. Eur. Y0000016, traceable to USP 1572503

vapor pressure

5.6 mmHg ( 100 °C)

API family

propofol

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.514 (lit.)

bp

256 °C/764 mmHg (lit.)

mp

18 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)c1cccc(C(C)C)c1O

InChI

1S/C12H18O/c1-8(2)10-6-5-7-11(9(3)4)12(10)13/h5-9,13H,1-4H3

InChI key

OLBCVFGFOZPWHH-UHFFFAOYSA-N

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Propofol is approved for induction and maintenance of paediatric anesthesia and sedation in children less than 3 years of age. The mechanism of action involves interaction with gamma-aminobutyric acid A receptor. Rapid effect and fewer side effects like postoperative nausea makes propofol a popular intravenous anesthetic agents.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Propofol may be used as a pharmaceutical reference standard for the determination of the analyte in emulsion dosage form by sequential injection technique with spectrophotometric and fluorimetric detections.

Analysis Note

These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
To see an example of a Certificate of Analysis for this material enter LRAA5999 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1572503BP1031Y0000016
Propofol British Pharmacopoeia (BP) Reference Standard

BP1031

Propofol

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

Quality Level

300

Quality Level

-

Quality Level

-

Quality Level

-

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Propofol


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Propofol: a review of its role in pediatric anesthesia and sedation
Chidambaran V, et al.
CNS Drugs, 29(7), 543-563 (2015)
Fully automated analytical procedure for propofol determination by sequential injection technique with spectrophotometric and fluorimetric detections
Sramkova I, et al.
Talanta, 118, 104-110 (2014)



Global Trade Item Number

SKUGTIN
PHR1663-1ML04061834801528

Questions

Reviews

No rating value

Active Filters