Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C11H10F3N3
CAS Number:
Molecular Weight:
241.21
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
grade
pharmaceutical primary standard
agency
USP
API family
nilotinib
packaging
pkg of 25 mg
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecules)
format
neat
storage temp.
2-8°C
SMILES string
NC1=CC(C(F)(F)F)=CC(N2C=NC(C)=C2)=C1
InChI
1S/C11H10F3N3/c1-7-5-17(6-16-7)10-3-8(11(12,13)14)2-9(15)4-10/h2-6H,15H2,1H3
InChI key
WWTGXYAJVXKEKL-UHFFFAOYSA-N
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Application
Nilotinib Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
1 of 1
This Item | |||
|---|---|---|---|
| application(s) pharmaceutical (small molecules) | application(s) pharmaceutical | application(s) pharmaceutical | application(s) pharmaceutical small molecule |
| format neat | format - | format neat | format - |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename USP | manufacturer/tradename - |
| packaging pkg of 25 mg | packaging pkg of 50 mg | packaging - | packaging pkg of 50 mg |
| API family nilotinib | API family nilotinib | API family nilotinib | API family nilotinib |
Still not finding the right product?
Explore all of our products under Nilotinib Related Compound A
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1463439-25MG | 04065265032629 |


