Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1463461

USP

Nilotinib System Suitability Mixture

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Methyl-N-[3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(3-pyridinyl)-2- pyrimidinyl]amino]-benzamide monohydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H22F3N7O · HCl · H2O
CAS Number:
Molecular Weight:
583.99
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

API family

nilotinib

packaging

pkg of 10 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecules)

format

neat

storage temp.

−20°C

SMILES string

FC(F)(F)c1cc(cc(c1)NC(=O)c3cc(c(cc3)C)Nc4nc(ccn4)c5cnccc5)[n]2cnc(c2)C.Cl.O

InChI

1S/C28H22F3N7O.ClH.H2O/c1-17-5-6-19(10-25(17)37-27-33-9-7-24(36-27)20-4-3-8-32-14-20)26(39)35-22-11-21(28(29,30)31)12-23(13-22)38-15-18(2)34-16-38;;/h3-16H,1-2H3,(H,35,39)(H,33,36,37);1H;1H2

InChI key

YCBPQSYLYYBPDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Nilotinib System Suitability Mixture USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service