Skip to Content
MilliporeSigma

1463457

USP

Nilotinib Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-Amino-4-methylbenzoic acid, 3-Amino-p-toluic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
H2NC6H3(CH3)CO2H
CAS Number:
Molecular Weight:
151.16
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
2082449
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

nilotinib

manufacturer/tradename

USP

mp

164-168 °C (lit.)

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1N)C(O)=O

InChI

1S/C8H9NO2/c1-5-2-3-6(8(10)11)4-7(5)9/h2-4H,9H2,1H3,(H,10,11)

InChI key

XKFIFYROMAAUDL-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nilotinib Related Compound C USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.


Still not finding the right product?

Explore all of our products under Nilotinib Related Compound C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library